4-Chlor-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chlor-2-fluorbenzolboronsäure, (4-Chlor-2-fluorphenyl)metaborsäure, Dihydroxy(4-chlor-2-fluorphenyl)boran

BB-2190-5G BB-2190-25G BB-2190-1G
COBBBB-2190-5GEA 53.1 EUR
COBBBB-2190-5G COBBBB-2190-25G COBBBB-2190-1G
4-Chlor-2-fluorphenylboronsäure
4-Chlor-2-fluorphenylboronsäure
Formel: C₆H₅BClFO₂
Molecular Weight: 174.37 g/mol
MDL: MFCD02684293
CAS-Nummer: 160591-91-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR