Methyl-4-chlor-2-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chlor-2-nitrobenzoesäuremethylester

CA-4537-5G CA-4537-25G CA-4537-100G
COBBCA-4537-5GEA 0 EUR
COBBCA-4537-5G COBBCA-4537-25G COBBCA-4537-100G
Methyl-4-chlor-2-nitrobenzoat
Methyl-4-chlor-2-nitrobenzoat
Formel: C₈H₆ClNO₄
Molecular Weight: 215,59 g/mol
MDL: MFCD00007213
CAS-Nummer: 42087-80-9
EINECS: 255-654-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR