4-Chlor-3-(trifluormethyl)benzolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 4-Chlor-3-(trifluormethyl)benzol-1-sulfonylchlorid, 4-Chlor-3-(trifluormethyl)benzolsulfonsäurechlorid, 4-Chlor-3-(trifluormethyl)benzolsulfochlorid

MO07002DA MO07002EA
MAYBMO07002DAEA 0 EUR
MAYBMO07002DA MAYBMO07002EA
4-Chlor-3-(trifluormethyl)benzolsulfonylchlorid
4-Chlor-3-(trifluormethyl)benzolsulfonylchlorid
Formel: C₇H₃Cl₂F₃O₂S
Molecular Weight: 279.07
MDL: MFCD00024882
CAS-Nummer: 32333-53-2
UN: 3261
ADR: 8 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR