4-Chlor-3-(trifluormethyl)benzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
4-Chlor-3-(trifluormethyl)benzol-1-sulfonylchlorid, 4-Chlor-3-(trifluormethyl)benzolsulfonsäurechlorid, 4-Chlor-3-(trifluormethyl)benzolsulfochlorid
013100-1G
013100-5G
FLUO013100-1GEA
36.9
EUR
FLUO013100-1G
FLUO013100-5G
4-Chlor-3-(trifluormethyl)benzolsulfonylchlorid
4-Chlor-3-(trifluormethyl)benzolsulfonylchlorid
Formel:
C₇H₃Cl₂F₃O₂S |
MDL:
MFCD00024882 CAS-Nummer: 32333-53-2 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...