Methyl-4-chlor-3-fluorbenzoat
Lieferant: Thermo Scientific
Synonyme:
4-Chlor-3-fluorbenzoesäuremethylester
RDP00212.250
RDP00212.1
MAYBRDP00212.250EA
0
EUR
MAYBRDP00212.250
MAYBRDP00212.1
Methyl-4-chlor-3-fluorbenzoat
Methyl-4-chlor-3-fluorbenzoat
Formel:
C₈H₆ClFO₂ Molecular Weight: 188.59 g/mol |
MDL:
MFCD00209299 CAS-Nummer: 206362-87-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...