4-Chlor-3-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Chlor-3-fluorphenyl)metaborsäure, Dihydroxy(4-chlor-3-fluorphenyl)boran, 4-Chlor-3-fluorbenzolboronsäure

BB-2256-5G BB-2256-25G
COBBBB-2256-5GEA 50.8 EUR
COBBBB-2256-5G COBBBB-2256-25G
4-Chlor-3-fluorphenylboronsäure
4-Chlor-3-fluorphenylboronsäure
Formel: C₆H₅BClFO₂
Molecular Weight: 174,37 g/mol
MDL: MFCD01319010
CAS-Nummer: 137504-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR