Methyl-4-chlor-3-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chlor-3-nitrobenzoesäuremethylester

CA-4683-25G CA-4683-100G CA-4683-5G
COBBCA-4683-25GEA 88.6 EUR
COBBCA-4683-25G COBBCA-4683-100G COBBCA-4683-5G
Methyl-4-chlor-3-nitrobenzoat
Methyl-4-chlor-3-nitrobenzoat
Formel: C₈H₆ClNO₄
Molecular Weight: 215,59 g/mol
MDL: MFCD00016989
CAS-Nummer: 14719-83-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR