4-Chlornicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chlor-3-pyridincarbonsäure, 4-Chlorpyridin-3-carbonsäure

PY-7247-100G PY-7247-25G PY-7247-5G
COBBPY-7247-100GEA 759 EUR
COBBPY-7247-100G COBBPY-7247-25G COBBPY-7247-5G
4-Chlornicotinsäure
4-Chlornicotinsäure
Formel: C₆H₄ClNO₂
Molecular Weight: 157,56 g/mol
MDL: MFCD00128860
CAS-Nummer: 10177-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR