4-Chlor-6-fluor-2-methylchinolin

Lieferant: COMBI-BLOCKS
Achtung

HC-2144-5G HC-2144-1G
COBBHC-2144-5GEA 451 EUR
COBBHC-2144-5G COBBHC-2144-1G
4-Chlor-6-fluor-2-methylchinolin
4-Chloro-6-fluoro-2-methylquinoline
Formel: C₁₀H₇ClFN
Molecular Weight: 195.62 g/mol
MDL: MFCD00272332
CAS-Nummer: 18529-01-6
EINECS: 242-403-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR