4-Chlorbenzamidin Hydrochlorid
Lieferant: Thermo Scientific
Synonyme:
4-Chlorbenzoesäureamidin Hydrochlorid, 4-Chlorphenylamidin Hydrochlorid
SPB06264DA
SPB06264EA
SPB06264.50
MAYBSPB06264DAEA
0
EUR
MAYBSPB06264DA
MAYBSPB06264EA
MAYBSPB06264.50
4-Chlorbenzamidin Hydrochlorid
4-Chlorbenzamidin Hydrochlorid
Formel:
C₇H₈Cl₂N₂ Molecular Weight: 191,06 g/mol |
MDL:
MFCD00126401 CAS-Nummer: 14401-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...