4-Chlorbenzamidin Hydrochlorid
Lieferant: CHEMPUR
Synonyme:
4-Chlorbenzoesäureamidin Hydrochlorid, 4-Chlorphenylamidin Hydrochlorid
12-0018.1
12-0018.5
CHMP12-0018.1EA
0
EUR
CHMP12-0018.1
CHMP12-0018.5
4-Chlorbenzamidin Hydrochlorid
4-Chlorbenzamidin Hydrochlorid
Formel:
C₇H₈Cl₂N₂ Molecular Weight: 191.06 g/mol |
MDL:
MFCD00126401 CAS-Nummer: 14401-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...