4-Chlorbenzamidin Hydroiodid 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Chlorbenzoesäureamidin Hydroiodid, 4-Chlorphenylamidin Hydroiodid

A11571.06 A11571.03
A11571.06EA 122 EUR
A11571.06 A11571.03
4-Chlorbenzamidin Hydroiodid 96%
4-Chlorbenzamidin Hydroiodid

Purity: 96% CAS No.: 115297-57-9 Molecular Formula: C7H7ClN2.HI Formula Weight: 282.51 Melting Point: ca 235° dec. Beilstein Reference: 8280383

Formel: C₇H₈ClIN₂
Molecular Weight: 282,51 g/mol
Schmelzpunkt: ∼ 235 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00051987
CAS-Nummer: 115297-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR