4-Chlorbenzhydrazid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chlorobenzoic hydrazide

HC-6821-5G HC-6821-25G
COBBHC-6821-5GEA 26.2 EUR
COBBHC-6821-5G COBBHC-6821-25G
4-Chlorbenzhydrazid
4-Chlorbenzhydrazid
Formel: C₇H₇ClN₂O
Molecular Weight: 170.6 g/mol
Siedepunkt: 354
Schmelzpunkt: 164
MDL: MFCD00007603
CAS-Nummer: 536-40-3
EINECS: 208-632-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR