4-Chlorphenylboronsäure
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
4-Chlorbenzolboronsäure, Dihydroxy(4-chlorphenyl)boran, (4-Chlorphenyl)metaborsäure
BB-2261-5G
BB-2261-25G
BB-2261-100G
COBBBB-2261-5GEA
25.5
EUR
COBBBB-2261-5G
COBBBB-2261-25G
COBBBB-2261-100G
4-Chlorphenylboronsäure
4-Chlorphenylboronsäure
Formel:
C₆H₆BClO₂ Molecular Weight: 156,38 g/mol Schmelzpunkt: 268…272 °C Dichte: ∼1 g/cm³ (20 °C) |
MDL:
MFCD00039137 CAS-Nummer: 1679-18-1 EINECS: 216-845-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...