4-Chlorthiobenzamid ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Chlorthiobenzoesäureamid

A12021.09 A12021.14 A12021.18 A12021.22
A12021.09EA 39.1 EUR
A12021.09 A12021.14 A12021.18 A12021.22
4-Chlorthiobenzamid ≥97%
4-Chlorthiobenzamid

Purity: 97% CAS No.: 2521-24-6 Molecular Formula: C7H6ClNS Formula Weight: 171.65 Melting Point: 123-129° Beilstein Reference: 606603

Formel: C₇H₆ClNS
Molecular Weight: 171,65 g/mol
Siedepunkt: 276 °C (1013 hPa)
Schmelzpunkt: 123…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040956
CAS-Nummer: 2521-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR