Indol-4-carbonitril
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
4-Indolcarbonitril, 4-Cyanindol, 1H-Indol-4-carbonitril, 4-Cyan-1H-indol
IN-0005-5G
IN-0005-25G
IN-0005-1G
COBBIN-0005-5GEA
37.9
EUR
COBBIN-0005-5G
COBBIN-0005-25G
COBBIN-0005-1G
Indol-4-carbonitril
Indol-4-carbonitril
Formel:
C₉H₆N₂ Molecular Weight: 142.16 g/mol |
MDL:
MFCD00152045 CAS-Nummer: 16136-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...