4-Cyanphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 4-Cyanobenzeneboronic acid, (4-Cyanophenyl)metaboric acid, Dihydroxy(4-cyanophenyl)borane

BB-2264-100G BB-2264-5G BB-2264-25G BB-2264-1G
COBBBB-2264-100GEA 398 EUR
COBBBB-2264-100G COBBBB-2264-5G COBBBB-2264-25G COBBBB-2264-1G
4-Cyanphenylboronsäure
4-Cyanphenylboronsäure
Formel: C₇H₆BNO₂
Molecular Weight: 146,94 g/mol
Schmelzpunkt: > 300 °C
MDL: MFCD01318968
CAS-Nummer: 126747-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR