4-Vinylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Dihydroxy(4-vinylphenyl)boran, (4-Vinylphenyl)metaborsäure, 4-Vinylbenzolboronsäure

BB-2676-5G BB-2676-25G BB-2676-1G
COBBBB-2676-5GEA 107 EUR
COBBBB-2676-5G COBBBB-2676-25G COBBBB-2676-1G
4-Vinylphenylboronsäure
4-Vinylphenylboronsäure
Formel: C₈H₉BO₂
Molecular Weight: 147,97 g/mol
MDL: MFCD00239441
CAS-Nummer: 2156-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR