4-Ethoxy-3-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2722-25G BB-2722-100G BB-2722-5G BB-2722-1G
COBBBB-2722-25GEA 277 EUR
COBBBB-2722-25G COBBBB-2722-100G COBBBB-2722-5G COBBBB-2722-1G
4-Ethoxy-3-fluorphenylboronsäure
4-Ethoxy-3-fluorphenylboronsäure
Formel: C₈H₁₀BFO₃
Molecular Weight: 183.97 g/mol
MDL: MFCD04115667
CAS-Nummer: 279263-10-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR