4-Ethoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Ethoxybenzeneboronic acid, (4-Ethoxyphenyl)metaboric acid, Dihydroxy(4-ethoxyphenyl)borane
BB-2628-5G
BB-2628-25G
COBBBB-2628-5GEA
38.5
EUR
COBBBB-2628-5G
COBBBB-2628-25G
4-Ethoxyphenylboronsäure
4-Ethoxyphenylboronsäure
Formel:
C₈H₁₁BO₃ Molecular Weight: 165,98 g/mol |
MDL:
MFCD00674028 CAS-Nummer: 22237-13-4 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...