4-Ethylbenzolsulfonylchlorid ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-Ethylbenzol-1-sulfonylchlorid, 4-Ethylbenzolsulfonsäurechlorid, 4-Ethylbenzolsulfochlorid

B24682.06 B24682.14
B24682.06EA 36.3 EUR
B24682.06 B24682.14
4-Ethylbenzolsulfonylchlorid ≥97%
4-Ethylbenzolsulfonylchlorid

Purity: 97% CAS No.: 16712-69-9 Molecular Formula: C8H9ClO2S Formula Weight: 204.68 Refractive Index: 1.5450 Melting Point: 8-12° Boiling Point: 134-136°/7mm Beilstein Reference: 2090130 UN No.: 3265 MDL No.: MFCD00041480

Formel: C₈H₉ClO₂S
Molecular Weight: 204,68 g/mol
Siedepunkt: 134…136 °C (7 mmHg)
Schmelzpunkt: 8…12 °C
Dichte: 1,268
Lagertemperatur: Raumtemperatur
MDL: MFCD00041480
CAS-Nummer: 16712-69-9
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR