4-Ethylphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: (4-Ethylphenyl)metaborsäure, 4-Ethylbenzolboronsäure, Dihydroxy(4-ethylphenyl)boran

BB-2670-100G BB-2670-5G BB-2670-25G
COBBBB-2670-100GEA 243 EUR
COBBBB-2670-100G COBBBB-2670-5G COBBBB-2670-25G
4-Ethylphenylboronsäure
4-Ethylphenylboronsäure
Formel: C₈H₁₁BO₂
Molecular Weight: 149.98 g/mol
Dichte: 1.125 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00859377
CAS-Nummer: 63139-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR