4-Fluor-o-anisaldehyd

Lieferant: FLUOROCHEM

Synonyme: 4-Fluor-2-anisaldehyd, 4-Fluor-2-methoxybenzaldehyd

013323-1G 013323-5G
FLUO013323-1GEA 39.9 EUR
FLUO013323-1G FLUO013323-5G
4-Fluor-o-anisaldehyd
4-Fluor-o-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00143318
CAS-Nummer: 450-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR