4-Fluor-2-nitrobenzaldehyd

Lieferant: COMBI-BLOCKS

OS-7634-5G OS-7634-25G OS-7634-1G
COBBOS-7634-5GEA 104 EUR
COBBOS-7634-5G COBBOS-7634-25G COBBOS-7634-1G
4-Fluor-2-nitrobenzaldehyd
4-Fluoro-2-nitrobenzaldehyde
Formel: C₇H₄FNO₃
MDL: MFCD01666439
CAS-Nummer: 2923-96-8
EINECS: 220-884-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR