4-Fluor-2-nitrobenzaldehyd

Lieferant: CHEMPUR

10-0064.1 10-0064.5
CHMP10-0064.1EA 0 EUR
CHMP10-0064.1 CHMP10-0064.5
4-Fluor-2-nitrobenzaldehyd
4-Fluoro-2-nitrobenzaldehyde
Formel: C₇H₄FNO₃
Molecular Weight: 169.11 g/mol
MDL: MFCD01666439
CAS-Nummer: 2923-96-8
EINECS: 220-884-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR