4-Fluor-2-nitrobenzoesäure

Lieferant: COMBI-BLOCKS

CA-4000-5G CA-4000-25G CA-4000-100G
COBBCA-4000-5GEA 0 EUR
COBBCA-4000-5G COBBCA-4000-25G COBBCA-4000-100G
4-Fluor-2-nitrobenzoesäure
4-Fluoro-2-nitrobenzoic acid
Formel: C₇H₄FNO₄
MDL: MFCD00024300
CAS-Nummer: 394-01-4
EINECS: 206-890-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR