Methyl-4-fluor-2-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluor-2-nitrobenzoesäuremethylester

CA-4464-5G CA-4464-1G
COBBCA-4464-5GEA 249 EUR
COBBCA-4464-5G COBBCA-4464-1G
Methyl-4-fluor-2-nitrobenzoat
Methyl-4-fluor-2-nitrobenzoat
Formel: C₈H₆FNO₄
Molecular Weight: 199,14 g/mol
MDL: MFCD09800921
CAS-Nummer: 151504-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR