4-Fluor-m-anisaldehyd

Lieferant: FLUOROCHEM

Synonyme: 4-Fluor-3-anisaldehyd, 4-Fluor-3-methoxybenzaldehyd

013012-5G 013012-25G
FLUO013012-5GEA 93.2 EUR
FLUO013012-5G FLUO013012-25G
4-Fluor-m-anisaldehyd
4-Fluor-m-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00143320
CAS-Nummer: 128495-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR