4-Fluor-m-anisaldehyd ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Fluor-3-anisaldehyd, 4-Fluor-3-methoxybenzaldehyd
L19671.03
L19671.06
L19671.03EA
24.5
EUR
L19671.03
L19671.06
4-Fluor-m-anisaldehyd ≥98%
4-Fluor-m-anisaldehyd
Purity: 98%
CAS No.: 128495-46-5
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Melting Point: 61-63°
Beilstein Reference: 5178063
Formel:
C₈H₇FO₂ Molecular Weight: 154,14 g/mol Schmelzpunkt: 61…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143320 CAS-Nummer: 128495-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...