4-Fluor-m-anisaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Fluor-3-anisaldehyd, 4-Fluor-3-methoxybenzaldehyd

M-Clarity™ Qualitätsstufe = MQ200

652687-25G 652687-5G
SIAL652687-25GEA 205 EUR
SIAL652687-25G SIAL652687-5G
4-Fluor-m-anisaldehyd, Sigma-Aldrich®
4-Fluor-m-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00143320
CAS-Nummer: 128495-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR