4-Fluor-m-toluylsäure

Lieferant: COMBI-BLOCKS

HC-6861-25G HC-6861-5G
COBBHC-6861-25GEA 88.6 EUR
COBBHC-6861-25G COBBHC-6861-5G
4-Fluor-m-toluylsäure
4-Fluor-m-toluylsäure
Formel: C₈H₇FO₂
Molecular Weight: 154.14 g/mol
MDL: MFCD01631424
CAS-Nummer: 403-15-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR