Methyl-4-fluor-3-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluor-3-nitrobenzoesäuremethylester

CA-4332-5G CA-4332-25G
COBBCA-4332-5GEA 0 EUR
COBBCA-4332-5G COBBCA-4332-25G
Methyl-4-fluor-3-nitrobenzoat
Methyl-4-fluor-3-nitrobenzoat
Formel: C₈H₆FNO₄
MDL: MFCD00196161
CAS-Nummer: 329-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR