5-Fluorisatin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
5-Fluor-1H-indol-2,3-dion, 5-Fluorindolin-2,3-dion, 5-Fluor-2,3-indolindion, 5-Fluor-2,3-dioxoindolin
IN-0144-25G
IN-0144-100G
COBBIN-0144-25GEA
89.2
EUR
COBBIN-0144-25G
COBBIN-0144-100G
5-Fluorisatin
5-Fluorisatin
Formel:
C₈H₄FNO₂ Molecular Weight: 165.12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022795 CAS-Nummer: 443-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...