4-Fluorbenzamidin Hydrochlorid

Lieferant: CHEMPUR

12-0036.1 12-0036.5
CHMP12-0036.1EA 0 EUR
CHMP12-0036.1 CHMP12-0036.5
4-Fluorbenzamidin Hydrochlorid
4-Fluorbenzamidin Hydrochlorid
Formel: C₇H₈ClFN₂
MDL: MFCD04114421
CAS-Nummer: 456-14-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR