4-Fluorbenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluorbenzolsulfonsäureamid

OT-0966-25G OT-0966-100G
COBBOT-0966-25GEA 111 EUR
COBBOT-0966-25G COBBOT-0966-100G
4-Fluorbenzolsulfonamid
4-Fluorbenzolsulfonamid
Formel: C₆H₆FNO₂S
Molecular Weight: 175,18 g/mol
MDL: MFCD00025384
CAS-Nummer: 402-46-0
EINECS: 206-946-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR