N,N-Dimethyl-4-fluorbenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluorbenzolsulfonsäuredimethylamid, N,N-Dimethyl-4-fluorbenzolsulfonamid

OT-0959-5G OT-0959-1G
COBBOT-0959-5GEA 310 EUR
COBBOT-0959-5G COBBOT-0959-1G
N,N-Dimethyl-4-fluorbenzolsulfonamid
N,N-Dimethyl-4-fluorbenzolsulfonamid
Formel: C₈H₁₀FNO₂S
Molecular Weight: 203,24 g/mol
MDL: MFCD01211990
CAS-Nummer: 383-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR