4-Fluorphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: Dihydroxy(4-fluorphenyl)boran, 4-Fluorbenzolboronsäure, (4-Fluorphenyl)metaborsäure

BB-2291-5G BB-2291-25G BB-2291-100G
COBBBB-2291-5GEA 26.2 EUR
COBBBB-2291-5G COBBBB-2291-25G COBBBB-2291-100G
4-Fluorphenylboronsäure
4-Fluorphenylboronsäure
Formel: C₆H₆BFO₂
Molecular Weight: 139,92 g/mol
Schmelzpunkt: 263…265 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039136
CAS-Nummer: 1765-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR