4-Hydroxybenzamid
Lieferant: FLUOROCHEM
Synonyme:
4-Hydroxybenzoesäureamid
002826-5G
002826-25G
FLUO002826-5GEA
36.9
EUR
FLUO002826-5G
FLUO002826-25G
4-Hydroxybenzamid
4-Hydroxybenzamid
Formel:
C₇H₇NO₂ Molecular Weight: 137,14 g/mol |
MDL:
MFCD00007997 CAS-Nummer: 619-57-8 EINECS: 210-602-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...