4-Hydroxybenzamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Hydroxybenzoesäureamid

M-Clarity™ Qualitätsstufe = MQ100

270253-25G 270253-5G
SIAL270253-25GEA 152 EUR
SIAL270253-25G SIAL270253-5G
4-Hydroxybenzamid, Sigma-Aldrich®
4-Hydroxybenzamid
Formel: C₇H₇NO₂
Molecular Weight: 137,14 g/mol
MDL: MFCD00007997
CAS-Nummer: 619-57-8
EINECS: 210-602-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR