4-Hydroxybenzamidin Hydrochlorid 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Hydroxyphenylamidin Hydrochlorid, 4-Hydroxybenzoesäureamidin Hydrochlorid

H31566.03 H31566.09
H31566.03EA 26.5 EUR
H31566.03 H31566.09
4-Hydroxybenzamidin Hydrochlorid 95%
4-Hydroxybenzamidin Hydrochlorid

Purity: 95% CAS No.: 38148-63-9 Molecular Formula: C7H8N2O.HCl Formula Weight: 172.61 Melting Point: 221-224° MDL No.: MFCD00136405

Formel: C₇H₉ClN₂O
Molecular Weight: 172,61 g/mol
Schmelzpunkt: 221…224 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00136405
CAS-Nummer: 38148-63-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR