4-Chinolinol
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
Quinolin-4-ol, 4-Hydroxyquinoline
HC-2717-25G
HC-2717-1G
HC-2717-5G
COBBHC-2717-25GEA
96.9
EUR
COBBHC-2717-25G
COBBHC-2717-1G
COBBHC-2717-5G
4-Chinolinol
4-Chinolinol
Formel:
C₉H₇NO Molecular Weight: 145.16 g/mol Schmelzpunkt: 202…206 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006777 CAS-Nummer: 611-36-9 EINECS: 210-268-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...