4-Hydroxyphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

BB-2152-25G BB-2152-1G BB-2152-5G BB-2152-100G
COBBBB-2152-25GEA 0 EUR
COBBBB-2152-25G COBBBB-2152-1G COBBBB-2152-5G COBBBB-2152-100G
4-Hydroxyphenylboronsäure
4-Hydroxyphenylboronsäure
Formel: C₆H₇BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01074628
CAS-Nummer: 71597-85-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR