4-Iod-2-methylimidazol

Lieferant: Thermo Scientific

Synonyme: 4-Iod-2-methyl-1H-imidazol

MO00831DA MO00831EA
MAYBMO00831DAEA 0 EUR
MAYBMO00831DA MAYBMO00831EA
4-Iod-2-methylimidazol
4-Iodo-2-methylimidazole
Formel: C₄H₅IN₂
Molecular Weight: 208 g/mol
MDL: MFCD02684335
CAS-Nummer: 73746-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR