4-Iodimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Iod-1H-imidazol

HI-1001-5G HI-1001-25G HI-1001-1G
COBBHI-1001-5GEA 61 EUR
COBBHI-1001-5G COBBHI-1001-25G COBBHI-1001-1G
4-Iodimidazol
4-Iodoimidazole
Formel: C₃H₃IN₂
Molecular Weight: 193,97 g/mol
Schmelzpunkt: 137…138 °C
MDL: MFCD01632213
CAS-Nummer: 71759-89-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR