4-Iodphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Jodbenzolboronsäure, 4-Iodbenzolboronsäure, (4-Iodphenyl)metaborsäure, (4-Jodphenyl)metaborsäure, Dihydroxy(4-jodphenyl)boran, Dihydroxy(4-iodphenyl)boran
BB-8335-5G
BB-8335-25G
BB-8335-1G
COBBBB-8335-5GEA
73.7
EUR
COBBBB-8335-5G
COBBBB-8335-25G
COBBBB-8335-1G
4-Iodphenylboronsäure
4-Iodphenylboronsäure
Formel:
C₆H₆BIO₂ Molecular Weight: 247.83 g/mol |
MDL:
MFCD01319014 CAS-Nummer: 5122-99-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...