4-Formylphenylboronsäure
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
(4-Formylphenyl)metaborsäure, 4-Boronobenzaldehyd, Dihydroxy(4-formylphenyl)boran, 4-Formylbenzolboronsäure
BB-2305-5G
BB-2305-25G
BB-2305-100G
COBBBB-2305-5GEA
26.2
EUR
COBBBB-2305-5G
COBBBB-2305-25G
COBBBB-2305-100G
4-Formylphenylboronsäure
4-Formylphenylboronsäure
Formel:
C₇H₇BO₃ Molecular Weight: 149,94 g/mol |
MDL:
MFCD00151823 CAS-Nummer: 87199-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...