4-Methoxybenzamid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Methoxybenzoesäureamid, p-Anisamid
B22614.09
B22614.18
B22614.09EA
55.4
EUR
B22614.09
B22614.18
4-Methoxybenzamid ≥98%
4-Methoxybenzamid
Purity: 98%
CAS No.: 3424-93-9
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Melting Point: 164-167°
Boiling Point: 295°
MDL No.: MFCD00007995
Formel:
C₈H₉NO₂ Molecular Weight: 151,16 g/mol Siedepunkt: 295 °C Schmelzpunkt: 164…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007995 CAS-Nummer: 3424-93-9 EINECS: 222-319-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...