4-Methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: (4-Methoxyphenyl)metaborsäure, Dihydroxy(4-methoxyphenyl)boran, 4-Methoxybenzolboronsäure

BB-2326-25G BB-2326-5G BB-2326-100G
COBBBB-2326-25GEA 50.9 EUR
COBBBB-2326-25G COBBBB-2326-5G COBBBB-2326-100G
4-Methoxyphenylboronsäure
4-Methoxyphenylboronsäure
Formel: C₇H₉BO₃
Molecular Weight: 151.96 g/mol
Schmelzpunkt: 204…207 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039139
CAS-Nummer: 5720-07-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR