4-Methyl-3-nitrobenzolsulfonylchlorid

Lieferant: CHEMPUR

Synonyme: 4-Methyl-3-nitrobenzol-1-sulfonylchlorid, 4-Methyl-3-nitrobenzolsulfonsäurechlorid, 4-Methyl-3-nitrobenzolsulfochlorid

FL13170.POR 013170-5G
CHMPFL13170.POREA 0 EUR
CHMPFL13170.POR FLUO013170-5G
4-Methyl-3-nitrobenzolsulfonylchlorid
4-Methyl-3-nitrobenzolsulfonylchlorid
Formel: C₇H₆ClNO₄S
Molecular Weight: 235.65
MDL: MFCD00129811
CAS-Nummer: 616-83-1
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR