4-Methyl-3-nitrobenzolsulfonylchlorid 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Methyl-3-nitrobenzol-1-sulfonylchlorid, 4-Methyl-3-nitrobenzolsulfonsäurechlorid, 4-Methyl-3-nitrobenzolsulfochlorid
A19931.06
A19931.22
A19931.06EA
69.9
EUR
A19931.06
A19931.22
4-Methyl-3-nitrobenzolsulfonylchlorid 95%
4-Methyl-3-nitrobenzolsulfonylchlorid
Purity: 95%
CAS No.: 616-83-1
Molecular Formula: C7H6ClNO4S
Formula Weight: 235.65
Melting Point: 33-36°
Boiling Point: 152-154°/1mm
UN No.: 3261
MDL No.: MFCD00129811
Formel:
C₇H₆ClNO₄S Molecular Weight: 235.65 Siedepunkt: 152…154 °C (1 mmHg) Schmelzpunkt: 33…36 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129811 CAS-Nummer: 616-83-1 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...